3-Fluoro-5-hydroxybenzoic acid 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD208889-25GEA 141 EUR
BLDPBD208889-25G BLDPBD208889-1G BLDPBD208889-100G BLDPBD208889-250MG BLDPBD208889-10G BLDPBD208889-5G
3-Fluoro-5-hydroxybenzoic acid 97%
3-Fluoro-5-hydroxybenzoic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₅FO₃
MW: 156,1112 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11846515
CAS-numero: 860296-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR