4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid 98%
|
|
BLDPBD212192-250MGEA
25
EUR
BLDPBD212192-250MG
BLDPBD212192-5G
BLDPBD212192-25G
BLDPBD212192-1G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₁₁H₁₇BN₂O₄ MW: 252,075 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD16660234 CAS-numero: 1083326-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...