3-tert-Butoxycarbonylamino-pentanoic acid 97%

BLDPBD212371-5GEA 608 EUR
BLDPBD212371-5G
3-tert-Butoxycarbonylamino-pentanoic acid 97%
3-tert-Butoxycarbonylamino-pentanoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₀H₁₉NO₄
MW: 217,262 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05863633
CAS-numero: 557091-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR