8-Bromo-6-methylimidazo[1,2-a]pyridine 98%

BLDPBD212567-5GEA 405 EUR
BLDPBD212567-5G BLDPBD212567-1G BLDPBD212567-250MG
8-Bromo-6-methylimidazo[1,2-a]pyridine 98%
8-Bromo-6-methylimidazo[1,2-a]pyridine

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₇BrN₂
MW: 211,0586 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09258770
CAS-numero: 136117-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR