3-Bromo-7-chloroquinoline 95+%

BLDPBD212632-1GEA 798 EUR
BLDPBD212632-1G
3-Bromo-7-chloroquinoline 95+%
3-Bromo-7-chloroquinoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₉H₅BrClN
MW: 242,5000 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18253966
CAS-numero: 84973-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR