3-Iodoanthranilic acid 97%

BLDPBD213030-100MGEA 17.7 EUR
BLDPBD213030-100MG BLDPBD213030-250MG BLDPBD213030-1G BLDPBD213030-25G BLDPBD213030-5G
3-Iodoanthranilic acid 97%
3-Iodoanthranilic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₇H₆INO₂
MW: 263,0300 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06808542
CAS-numero: 20776-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR