5-Iodo-o-toluic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD21387-25GEA 15.6 EUR
BLDPBD21387-25G BLDPBD21387-100G BLDPBD21387-10G BLDPBD21387-500G BLDPBD21387-5G
5-Iodo-o-toluic acid 98%
5-Iodo-o-toluic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₈H₇IO₂
MW: 262,0400 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01570258
CAS-numero: 54811-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR