4,7-Dibromo-2,1,3-benzoxadiazole 97%

BLDPBD215455-100MGEA 73.8 EUR
BLDPBD215455-100MG BLDPBD215455-250MG BLDPBD215455-25G BLDPBD215455-0.025 BLDPBD215455-10G BLDPBD215455-5G BLDPBD215455-1G
4,7-Dibromo-2,1,3-benzoxadiazole 97%
4,7-Dibromo-2,1,3-benzoxadiazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₆H₂Br₂N₂O
MW: 277,90 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02114623
CAS-numero: 54286-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR