6-Cyclopropylpyridin-3-amine 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD216795-100MGEA 56.2 EUR
BLDPBD216795-100MG BLDPBD216795-5G BLDPBD216795-1G BLDPBD216795-250MG
6-Cyclopropylpyridin-3-amine 95%
6-Cyclopropylpyridin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD12033370
CAS-numero: 1159821-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR