1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine 98%

BLDPBD217234-25GEA 67.6 EUR
BLDPBD217234-25G BLDPBD217234-5G BLDPBD217234-10G BLDPBD217234-1G BLDPBD217234-100G
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine 98%
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₀H₁₄N₂O₂
MW: 194,23 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD12761241
CAS-numero: 1153949-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR