3-Bromo-6-chloro-2-fluorobenzaldehyde 98%
Toimittaja: BLD PHARMATECH GMBH
|
|
BLDPBD217974-1GEA
75.9
EUR
BLDPBD217974-1G
BLDPBD217974-250MG
BLDPBD217974-5G
3-Bromo-6-chloro-2-fluorobenzaldehyde 98%
3-Bromo-6-chloro-2-fluorobenzaldehyde
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Kaava:
C₇H₃BrClFO MW: 237,4500 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD11111905 CAS-numero: 886615-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...