2,6-Difluoro-3-iodopyridine 95%

BLDPBD217997-25GEA 169 EUR
BLDPBD217997-25G BLDPBD217997-5G BLDPBD217997-100G BLDPBD217997-1G BLDPBD217997-10G BLDPBD217997-0.025
2,6-Difluoro-3-iodopyridine 95%
2,6-Difluoro-3-iodopyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₅H₂F₂IN
MW: 240,98 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09743595
CAS-numero: 685517-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR