6-Bromoimidazo[1,2-a]pyridine 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD21819-500GEA 817 EUR
BLDPBD21819-500G BLDPBD21819-5G BLDPBD21819-10G BLDPBD21819-1G BLDPBD21819-100G BLDPBD21819-25G
6-Bromoimidazo[1,2-a]pyridine 97%
6-Bromoimidazo[1,2-a]pyridine

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₅BrN₂
MW: 197,03 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06496200
CAS-numero: 6188-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR