4-(trans-2-Carboxyvinyl)phenylboronic acid 96%

BLDPBD218882-1GEA 74.9 EUR
BLDPBD218882-1G BLDPBD218882-5G BLDPBD218882-250MG
4-(trans-2-Carboxyvinyl)phenylboronic acid 96%
4-(trans-2-Carboxyvinyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₉H₉BO₄
MW: 191,9764 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD01075704
CAS-numero: 159896-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR