Methyl 3-aminobenzofuran-2-carboxylate 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD218896-250MGEA 33.3 EUR
BLDPBD218896-250MG BLDPBD218896-5G BLDPBD218896-1G
Methyl 3-aminobenzofuran-2-carboxylate 98%
Methyl 3-aminobenzofuran-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₁₀H₉NO₃
MW: 191,183 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00466275
CAS-numero: 57805-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR