4-Bromo-2-fluorobenzamide 96%

BLDPBD219467-250MGEA 6.3 EUR
BLDPBD219467-250MG BLDPBD219467-100G BLDPBD219467-5G BLDPBD219467-25G BLDPBD219467-1G
4-Bromo-2-fluorobenzamide 96%
4-Bromo-2-fluorobenzamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₇H₅BrFNO
MW: 218,03 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03094432
CAS-numero: 292621-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR