4-Methyl-5-nitro-1H-indazole 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD219869-100MGEA 27.1 EUR
BLDPBD219869-100MG BLDPBD219869-250MG BLDPBD219869-5G BLDPBD219869-1G
4-Methyl-5-nitro-1H-indazole 95%
4-Methyl-5-nitro-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Kaava: C₈H₇N₃O₂
MW: 177,1600 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03787902
CAS-numero: 101420-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR