2-Chloro-5-(trifluoromethyl)benzyl alcohol 95%

BLDPBD21993-5GEA 17.7 EUR
BLDPBD21993-5G BLDPBD21993-100G BLDPBD21993-25G
2-Chloro-5-(trifluoromethyl)benzyl alcohol 95%
2-Chloro-5-(trifluoromethyl)benzyl alcohol

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₆ClF₃O
MW: 210,58 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01631469
CAS-numero: 64372-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR