Chelidamic acid monohydrate 98%

BLDPBD220061-5GEA 21.9 EUR
BLDPBD220061-5G BLDPBD220061-10G BLDPBD220061-100G BLDPBD220061-25G BLDPBD220061-0.025 BLDPBD220061-1G
Chelidamic acid monohydrate 98%
Chelidamic acid monohydrate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₅NO₅·H₂O
MW: 201,134 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03818117
CAS-numero: 199926-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR