N,N-Dimethyl 3-boronobenzenesulphonamide 98%

BLDPBD221148-100MGEA 40.6 EUR
BLDPBD221148-100MG
N,N-Dimethyl 3-boronobenzenesulphonamide 98%
N,N-Dimethyl 3-boronobenzenesulphonamide

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₈H₁₂BNO₄S
MW: 229,06 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07363747
CAS-numero: 871329-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR