8-Amino-6-bromoquinoline 95%

BLDPBD223733-250MGEA 83.2 EUR
BLDPBD223733-250MG BLDPBD223733-1G BLDPBD223733-100MG
8-Amino-6-bromoquinoline 95%
8-Amino-6-bromoquinoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₉H₇BrN₂
MW: 223,0700 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00194777
CAS-numero: 57339-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR