Moxonidine 98%

BLDPBD22498-5GEA 179 EUR
BLDPBD22498-5G BLDPBD22498-250MG BLDPBD22498-1G BLDPBD22498-100MG
Moxonidine 98%
Moxonidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₉H₁₂ClN₅O
MW: 241,68 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00865794
CAS-numero: 75438-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR