2-Butynoic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD2250-1GEA 7.3 EUR
BLDPBD2250-1G BLDPBD2250-10G BLDPBD2250-5G BLDPBD2250-25G
2-Butynoic acid 98%
2-Butynoic acid

Storage condition: Sealed in dry, 2-8°C

Kaava: C₄H₄O₂
MW: 84,07 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00004363
CAS-numero: 590-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR