5-Pyrimidineboronic acid 98%
|
|
BLDPBD2257-10GEA
57.2
EUR
BLDPBD2257-10G
BLDPBD2257-1G
BLDPBD2257-25G
BLDPBD2257-500G
BLDPBD2257-100G
BLDPBD2257-5G
5-Pyrimidineboronic acid 98%
5-Pyrimidineboronic acid
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₄H₅BN₂O₂ MW: 123,91 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD03002366 CAS-numero: 109299-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...