2-Bromomaleimide 97%

BLDPBD226511-1GEA 101 EUR
BLDPBD226511-1G BLDPBD226511-250MG BLDPBD226511-5G BLDPBD226511-100MG
2-Bromomaleimide 97%
2-Bromomaleimide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₄H₂BrNO₂
MW: 175,968 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09035063
CAS-numero: 98026-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR