5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD227477-1GEA 180 EUR
BLDPBD227477-1G BLDPBD227477-250MG BLDPBD227477-5G BLDPBD227477-100MG BLDPBD227477-25G
5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected 98%
5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₇H₂₆BNO₆
MW: 351,202 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD11855974
CAS-numero: 942070-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR