4-(Cyanomethoxy)phenylboronic acid pinacol ester 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD228667-5GEA 215 EUR
BLDPBD228667-5G BLDPBD228667-1G BLDPBD228667-250MG
4-(Cyanomethoxy)phenylboronic acid pinacol ester 98%
4-(Cyanomethoxy)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₄H₁₈BNO₃
MW: 259,1100 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06795675
CAS-numero: 475272-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR