1-Boc-5-methoxyindole-3-boronic acid pinacol ester 98%

BLDPBD230096-1GEA 183 EUR
BLDPBD230096-1G BLDPBD230096-5G BLDPBD230096-250MG BLDPBD230096-25G BLDPBD230096-100MG
1-Boc-5-methoxyindole-3-boronic acid pinacol ester 98%
1-Boc-5-methoxyindole-3-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₂₀H₂₈BNO₅
MW: 373,2510 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD16295115
CAS-numero: 1256359-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR