3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine 98%

BLDPBD230755-1GEA 71.7 EUR
BLDPBD230755-1G BLDPBD230755-250MG BLDPBD230755-5G BLDPBD230755-10G
3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine 98%
3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₈H₇ClN₄
MW: 194,6210 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09800928
CAS-numero: 941294-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR