5-Chloro-(2-methoxycarbonyl)phenylboronic acid 98%

BLDPBD231351-250MGEA 48.9 EUR
BLDPBD231351-250MG BLDPBD231351-1G BLDPBD231351-25G BLDPBD231351-5G
5-Chloro-(2-methoxycarbonyl)phenylboronic acid 98%
5-Chloro-(2-methoxycarbonyl)phenylboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₈H₈BClO₄
MW: 214,4100 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD09878336
CAS-numero: 957062-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR