3-(Cyanomethoxy)phenylboronic acid 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD231471-250MGEA 68.6 EUR
BLDPBD231471-250MG BLDPBD231471-100MG BLDPBD231471-1G
3-(Cyanomethoxy)phenylboronic acid 95%
3-(Cyanomethoxy)phenylboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₈H₈BNO₃
MW: 176,9700 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD09908169
CAS-numero: 947533-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR