5-Fluoro-2-hydroxybenzonitrile 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD232059-100MGEA 21.9 EUR
BLDPBD232059-100MG BLDPBD232059-5G BLDPBD232059-1G BLDPBD232059-250MG
5-Fluoro-2-hydroxybenzonitrile 98%
5-Fluoro-2-hydroxybenzonitrile

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₄FNO
MW: 137,1100 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11846513
CAS-numero: 91407-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR