2-Aminofuran, N-BOC protected 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD232409-1GEA 35.4 EUR
BLDPBD232409-1G BLDPBD232409-5G BLDPBD232409-0.025 BLDPBD232409-25G BLDPBD232409-250MG
2-Aminofuran, N-BOC protected 97%
2-Aminofuran, N-BOC protected

Storage condition: Sealed in dry, 2-8°C

Kaava: C₉H₁₃NO₃
MW: 183,204 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD02682023
CAS-numero: 56267-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR