[(4-Methoxyphenyl)ethynyl]trimethylsilane 95+%
|
|
BLDPBD233483-5GEA
211
EUR
BLDPBD233483-5G
BLDPBD233483-250MG
BLDPBD233483-1G
BLDPBD233483-25G
[(4-Methoxyphenyl)ethynyl]trimethylsilane 95+%
[(4-Methoxyphenyl)ethynyl]trimethylsilane
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₂H₁₆OSi MW: 204,34 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD04039887 CAS-numero: 3989-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...