2,2-Dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD233541-250MGEA 18.7 EUR
BLDPBD233541-250MG BLDPBD233541-1G BLDPBD233541-5G
2,2-Dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one 97%
2,2-Dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₉N₃O₄
MW: 223,185 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD10000811
CAS-numero: 1002726-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR