7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%

BLDPBD234355-1GEA 192 EUR
BLDPBD234355-1G BLDPBD234355-250MG BLDPBD234355-100MG BLDPBD234355-5G
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₇H₅N₃O
MW: 147,134 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD13189361
CAS-numero: 1060815-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR