Methyl 5-bromo-1H-indazole-4-carboxylate 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD235419-5GEA 1760 EUR
BLDPBD235419-5G BLDPBD235419-250MG BLDPBD235419-100MG BLDPBD235419-1G
Methyl 5-bromo-1H-indazole-4-carboxylate 95%
Methyl 5-bromo-1H-indazole-4-carboxylate

Storage condition: Sealed in dry, 2-8°C

Kaava: C₉H₇BrN₂O₂
MW: 255,068 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD13177017
CAS-numero: 1037840-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR