6-Iodo-2,3-dihydro-1H-isoindol-1-one 95%

BLDPBD235525-100MGEA 85.2 EUR
BLDPBD235525-100MG BLDPBD235525-1G BLDPBD235525-250MG
6-Iodo-2,3-dihydro-1H-isoindol-1-one 95%
6-Iodo-2,3-dihydro-1H-isoindol-1-one

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₈H₆INO
MW: 259,044 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13177830
CAS-numero: 675109-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR