3-(Diisopropylcarbamoyl)phenylboronic acid 98%

BLDPBD236278-250MGEA 31.2 EUR
BLDPBD236278-250MG BLDPBD236278-5G BLDPBD236278-1G
3-(Diisopropylcarbamoyl)phenylboronic acid 98%
3-(Diisopropylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₃H₂₀BNO₃
MW: 249,1138 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06659890
CAS-numero: 850567-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR