5,6-Dimethyl-1-indanone 96%

BLDPBD237489-1GEA 64.5 EUR
BLDPBD237489-1G BLDPBD237489-25G BLDPBD237489-5G BLDPBD237489-250MG
5,6-Dimethyl-1-indanone 96%
5,6-Dimethyl-1-indanone

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₁H₁₂O
MW: 160,212 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00666126
CAS-numero: 16440-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR