2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one 98%

BLDPBD237568-5GEA 34.3 EUR
BLDPBD237568-5G BLDPBD237568-25G BLDPBD237568-100G
2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one 98%
2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₄H₉NO₃
MW: 239,23 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD02234511
CAS-numero: 1218-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR