2,7-Diiodo-9,9-dimethylfluorene 98%

BLDPBD238722-1GEA 56.2 EUR
BLDPBD238722-1G BLDPBD238722-250MG BLDPBD238722-5G
2,7-Diiodo-9,9-dimethylfluorene 98%
2,7-Diiodo-9,9-dimethylfluorene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₅H₁₂I₂
MW: 446,07 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD12827985
CAS-numero: 144981-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR