2,6-Diethynylpyridine 98%

BLDPBD239418-1GEA 83.2 EUR
BLDPBD239418-1G BLDPBD239418-250MG BLDPBD239418-5G
2,6-Diethynylpyridine 98%
2,6-Diethynylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₉H₅N
MW: 127,15 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09039127
CAS-numero: 75867-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR