2-Amino-5-nitrobenzamide 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD241315-250MGEA 81.1 EUR
BLDPBD241315-250MG
2-Amino-5-nitrobenzamide 95%
2-Amino-5-nitrobenzamide
Kaava: C₇H₇N₃O₃
MW: 181,149 g/mol
MDL numero: MFCD00090235
CAS-numero: 16313-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR