2-[2-(2-Aminoethoxy)ethoxy]acetic acid 95%

BLDPBD242056-5GEA 126 EUR
BLDPBD242056-5G BLDPBD242056-250MG BLDPBD242056-25G BLDPBD242056-1G BLDPBD242056-10G
2-[2-(2-Aminoethoxy)ethoxy]acetic acid 95%
2-[2-(2-Aminoethoxy)ethoxy]acetic acid

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Kaava: C₆H₁₃NO₄
MW: 163,17 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD13185897
CAS-numero: 134978-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR