Methyl hydrazinecarbodithioate 95%

BLDPBD242444-1GEA 40.6 EUR
BLDPBD242444-1G BLDPBD242444-25G BLDPBD242444-5G
Methyl hydrazinecarbodithioate 95%
Methyl hydrazinecarbodithioate
Kaava: C₂H₆N₂S₂
MW: 122,212 g/mol
MDL numero: MFCD00463356
CAS-numero: 5397-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR