Benzo[b]benzo[4,5]thieno[2,3-d]thiophene 97%

BLDPBD242735-10GEA 286 EUR
BLDPBD242735-10G BLDPBD242735-25G BLDPBD242735-1G BLDPBD242735-5G BLDPBD242735-250MG BLDPBD242735-100MG
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene 97%
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₄H₈S₂
MW: 240,34 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02114004
CAS-numero: 248-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR