4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide 97%

BLDPBD244423-5GEA 80 EUR
BLDPBD244423-5G BLDPBD244423-10G BLDPBD244423-1G BLDPBD244423-25G BLDPBD244423-250MG
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide 97%
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₃H₁₂FN₃O₂
MW: 261,26 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09033852
CAS-numero: 757251-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR