3,6-Dibromo-9-(p-tolyl)-9H-carbazole 98%
|
|
BLDPBD246836-1GEA
167
EUR
BLDPBD246836-1G
BLDPBD246836-5G
BLDPBD246836-250MG
3,6-Dibromo-9-(p-tolyl)-9H-carbazole 98%
3,6-Dibromo-9-(p-tolyl)-9H-carbazole
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₉H₁₃Br₂N MW: 415,13 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD12024282 CAS-numero: 357437-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...