1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD246900-250MGEA 22.9 EUR
BLDPBD246900-250MG BLDPBD246900-1G BLDPBD246900-5G
1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid 98%
1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₉H₇N₃O₂
MW: 189,171 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18260889
CAS-numero: 1014631-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR